2-Bromo-5-methoxypyridine 98%

Supplier: Ambeed
Warning

A226003-100G A226003-10G A226003-1G A226003-25G A226003-5G
103819-210EA 11 USD
103819-210 103850-112 77000-064 77000-066 77088-834 77088-836 77088-838
2-Bromo-5-methoxypyridine 98%
2-Bromo-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 240.8 °C (760 mm Hg)
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD07781221
CAS Number: 105170-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR