3-Amino-2-bromo-5-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-methoxypyridin-3-amine

A127812-10G A127812-250MG A127812-50MG A127812-5G
103801-016EA 46 USD
103801-016 76782-130 76782-132 76782-134 77313-282
3-Amino-2-bromo-5-methoxypyridine 98%
3-Amino-2-bromo-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.0366 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16659663
CAS Number: 1043688-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR