2-Bromo-5-methoxyphenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Brom-3-hydroxyanisole

A246233-1G A246233-25G A246233-5G A246233-100G A246233-10G A246233-250MG
103811-694EA 7 USD
103811-694 103851-522 77090-808 77090-810 77090-812 77261-910 77261-912 77261-914
2-Bromo-5-methoxyphenol 98%
2-Bromo-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
Boiling Pt: 253.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03428523
CAS Number: 63604-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR