2-Bromo-5-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A115245-100G A115245-10G A115245-25G A115245-500G A115245-5G
103824-384EA 5 USD
103824-384 77233-866 77233-868 77233-870 77233-872 77297-354
2-Bromo-5-methoxybenzoic acid 98%
2-Bromo-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Boiling Pt: 337.8 °C (760 mm Hg)
Melting Pt: 157…159 °C
Flash Pt: 158.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00020214
CAS Number: 22921-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR