6-Bromo-m-anisaldehyde 98%

Supplier: Ambeed
Warning

A218152-1G A218152-25G A218152-5G A218152-100G A218152-10G A218152-500G
103818-698EA 9 USD
103818-698 103849-426 77088-084 77088-086 77088-088 77237-590 77237-592 77237-594
6-Bromo-m-anisaldehyde 98%
6-Bromo-m-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Melting Pt: 71…76 °C
Flash Pt: 125.515 °C
Storage Temperature: Ambient
MDL Number: MFCD00463817
CAS Number: 7507-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR