2-Bromo-5-methoxyaniline 98%
Supplier: Ambeed
|
Warning
|
A212823-25G
A212823-5G
A212823-100G
A212823-10G
A212823-500G
A212823-1G
103818-242EA
6
USD
103818-242
103848-938
77087-414
77087-416
77165-006
77165-008
77165-010
77303-262
2-Bromo-5-methoxyaniline 98%
2-Bromo-5-methoxyaniline
Precaución: for Research Use Only
Formula:
C₇H₈BrNO Flash Pt: 123.173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070741 CAS Number: 59557-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...