2-Bromo-5-isopropylthiazole 97%

Supplier: Ambeed
Warning

A184181-5G A184181-250MG
103815-724EA 62 USD
103815-724 103846-286 77301-886 77881-388
2-Bromo-5-isopropylthiazole 97%
2-Bromo-5-isopropylthiazole

Precaución: for Research Use Only

Formula: C₆H₈BrNS
Storage Temperature: Refrigerator
MDL Number: MFCD12033480
CAS Number: 1159817-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR