2-Bromo-5-iodopyridine 97%

Supplier: Ambeed
Warning

A126780-100G A126780-10G A126780-500G A126780-1G A126780-25G A126780-5G
103800-862EA 9 USD
103800-862 76810-958 76810-960 76810-962 77298-150 77313-092 77313-094
2-Bromo-5-iodopyridine 97%
2-Bromo-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
Melting Pt: 121…123 °C
Flash Pt: 122.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03095201
CAS Number: 73290-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR