2-Bromo-5-iodopyrazine 97%

Supplier: Ambeed
Warning

A311500-1G A311500-25G A311500-5G A311500-100G A311500-10G A311500-250MG
103814-668EA 13 USD
103814-668 103835-858 76838-658 76838-660 76838-662 77263-218 77263-220 77263-222
2-Bromo-5-iodopyrazine 97%
2-Bromo-5-iodopyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrIN₂
MW: 284.8800 g/mol
Melting Pt: 95…97 °C
Flash Pt: 119.335 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061602
CAS Number: 622392-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR