2-Bromo-5-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A324637-100MG A324637-10G A324637-250MG A324637-25G A324637-1G A324637-5G
103825-372EA 23 USD
103825-372 103836-316 76828-240 76828-242 76828-244 76828-246 76839-236 77306-316
2-Bromo-5-iodobenzonitrile 98%
2-Bromo-5-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
Storage Temperature: Ambient
MDL Number: MFCD09025663
CAS Number: 1252046-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR