2-Bromo-5-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A201001-100G A201001-10G A201001-1G A201001-25G A201001-5G
103817-260EA 9 USD
103817-260 103847-896 76770-144 76770-146 77085-966 77085-968 77085-970
2-Bromo-5-iodobenzoic acid 98%
2-Bromo-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Boiling Pt: 371.2 °C (760 mm Hg)
Melting Pt: 171…175 °C
Flash Pt: 178.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00079716
CAS Number: 25252-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR