2-Bromo-5-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A215119-10G A215119-250MG A215119-1G A215119-5G
103818-454EA 23 USD
103818-454 103849-158 76814-694 76814-696 77087-682 77087-684
2-Bromo-5-iodobenzaldehyde 97%
2-Bromo-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD07779027
CAS Number: 1032231-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR