2-Bromopyrimidin-5-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-hydroxypyrimidine

A307975-250MG A307975-1G A307975-10G
103814-542EA 18 USD
103814-542 103835-718 76816-864 76838-474 77672-562
2-Bromopyrimidin-5-ol 97%
2-Bromopyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O
MW: 174.983 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09953508
CAS Number: 1240621-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR