2-Bromo-5-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A278873-1G A278873-5G A278873-10G A278873-250MG A278873-25G
103813-432EA 14 USD
103813-432 103853-376 77093-300 77093-302 77262-596 77262-598 77305-306
2-Bromo-5-hydroxybenzonitrile 98%
2-Bromo-5-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrNO
MW: 198.0200 g/mol
Boiling Pt: 321.2 °C (760 mm Hg)
Flash Pt: 148.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08061922
CAS Number: 189680-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR