2-Bromo-5-hydroxybenzoic acid 96%

Supplier: Ambeed
Warning

A158446-10G A158446-250MG A158446-1G A158446-25G A158446-5G
103805-130EA 7 USD
103805-130 103843-340 77028-524 77028-526 77319-198 77319-200 77319-202
2-Bromo-5-hydroxybenzoic acid 96%
2-Bromo-5-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₃
Boiling Pt: 374.5 °C (760 mm Hg)
Flash Pt: 180.3 °C
Storage Temperature: Ambient
MDL Number: MFCD06797974
CAS Number: 58380-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR