2-Bromo-5-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Danger
|
A182047-25G
A182047-5G
A182047-10G
A182047-1G
A182047-100G
103807-682EA
7
USD
103807-682
103846-050
77083-582
77083-584
77138-758
77301-766
77615-532
2-Bromo-5-hydroxybenzaldehyde 97%
2-Bromo-5-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol Boiling Pt: 286.7 °C (760 mm Hg) Melting Pt: 130…135 °C Flash Pt: 127.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03033065 CAS Number: 2973-80-0 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...