2-Bromo-5-hydroxy-4-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A214489-100G A214489-10G A214489-1G A214489-25G A214489-5G
103818-408EA 6 USD
103818-408 103849-114 76825-976 76825-978 77087-634 77087-636 77087-638
2-Bromo-5-hydroxy-4-methoxybenzoic acid 98%
2-Bromo-5-hydroxy-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₄
MW: 247.043 g/mol
Boiling Pt: 367 °C (760 mm Hg)
Melting Pt: 165…169 °C
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD16659606
CAS Number: 121936-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR