2-Bromo-5H-pyrrolo[2,3-b]pyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4,7-diazaindole

A112243-100G A112243-10G A112243-25G A112243-1G
103797-986EA 14 USD
103797-986 103823-822 77148-754 77148-756 77148-758 77310-624
2-Bromo-5H-pyrrolo[2,3-b]pyrazine 98%
2-Bromo-5H-pyrrolo[2,3-b]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
Boiling Pt: 265 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834822
CAS Number: 875781-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR