2-Bromo-5-formylthiazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromothiazole-5-carboxaldehyde

A242516-250MG A242516-1G A242516-5G A242516-10G
103811-472EA 11 USD
103811-472 103851-284 76815-462 77090-458 77090-460 77568-356
2-Bromo-5-formylthiazole 98%
2-Bromo-5-formylthiazole

Precaución: For research use only.

Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Boiling Pt: 264.9 °C (760 mm Hg)
Melting Pt: 94…96 °C
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788567
CAS Number: 464192-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR