2-Bromo-5-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A335661-25G
A335661-5G
A335661-100G
A335661-10G
A335661-500G
A335661-1G
103825-698EA
5
USD
103825-698
103836-670
76839-682
76839-684
77277-886
77277-888
77277-890
77306-492
2-Bromo-5-fluoropyridine 98%
2-Bromo-5-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₃BrFN Boiling Pt: 171.6 °C (760 mm Hg) Melting Pt: 30…31 °C Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234011 CAS Number: 41404-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...