2-Bromo-5-fluoropyrazine 98%

Supplier: Ambeed
Warning

A191538-100MG A191538-250MG A191538-5G A191538-1G
103816-382EA 111 USD
103816-382 77012-348 77012-350 77012-352 77084-822
2-Bromo-5-fluoropyrazine 98%
2-Bromo-5-fluoropyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrFN₂
Boiling Pt: 176 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD14702726
CAS Number: 1209459-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR