(2-Bromo-5-fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-fluorophenylacetic acid

A158539-10G A158539-250MG A158539-25G A158539-1G A158539-5G
103805-138EA 14 USD
103805-138 103843-350 77028-548 77028-550 77300-336 77319-214 77319-216
(2-Bromo-5-fluorophenyl)acetic acid 98%
(2-Bromo-5-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Boiling Pt: 310.7±27.0 °C (760 mm Hg)
Melting Pt: 113…114 °C
Flash Pt: 141.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03094440
CAS Number: 739336-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR