2-Bromo-5-fluorophenol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Bromo-4-fluoro-2-hydroxybenzene
A568552-25G
A568552-5G
A568552-100G
A568552-10G
A568552-1G
A568552-500G
103839-938EA
13
USD
103839-938
76844-066
76844-068
77292-594
77292-596
77292-598
77292-600
2-Bromo-5-fluorophenol 97%
2-Bromo-5-fluorophenol
Precaución: for Research Use Only
Formula:
C₆H₄BrFO Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040939 CAS Number: 147460-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...