1-Bromo-4-fluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A306456-25G A306456-5G A306456-100G A306456-10G
103835-632EA 7 USD
103835-632 76838-338 76838-340 77290-106 77290-108
1-Bromo-4-fluoro-2-nitrobenzene 98%
1-Bromo-4-fluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
Boiling Pt: 220.9 °C (760 mm Hg)
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00055530
CAS Number: 446-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR