2-Bromo-5-fluoroisonicotinaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-fluoro-4-formylpyridine, 2-Bromo-5-fluoropyridine-4-carboxaldehyde
A257336-250MG
A257336-5G
A257336-1G
103812-298EA
24
USD
103812-298
76976-844
76976-846
77091-676
2-Bromo-5-fluoroisonicotinaldehyde 95%
2-Bromo-5-fluoroisonicotinaldehyde
Precaución: for Research Use Only
Formula:
C₆H₃BrFNO MW: 203.997 g/mol Boiling Pt: 251.7±35.0 °C (760 mm Hg) Flash Pt: 106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09800558 CAS Number: 1005291-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...