2-Bromo-5-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A377282-25G A377282-100G A377282-10G A377282-500G
103837-636EA 8 USD
103837-636 76840-944 77115-460 77115-462 77115-464
2-Bromo-5-fluorobenzoic acid 98%
2-Bromo-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Boiling Pt: 291.1 °C (760 mm Hg)
Melting Pt: 154…157 °C
Flash Pt: 129.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR