2-Bromo-5-fluoro-6-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A113719-250MG
A113719-5G
A113719-100G
A113719-25G
103798-276EA
14
USD
103798-276
103824-118
77094-116
77310-930
77549-214
77549-216
2-Bromo-5-fluoro-6-methylpyridine 98%
2-Bromo-5-fluoro-6-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 183.4 °C (760 mm Hg) Melting Pt: 63…65 °C Flash Pt: 64.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03095064 CAS Number: 374633-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...