2-Bromo-5-fluoroisonicotinic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-fluoro-4-pyridinecarboxylic acid, 2-Bromo-5-fluoropyridine-4-carboxylic acid
A238006-10G
A238006-250MG
A238006-1G
A238006-5G
A238006-25G
A238006-100G
103811-192EA
10
USD
103811-192
103850-976
77031-214
77031-216
77090-070
77090-072
77304-098
77672-128
2-Bromo-5-fluoroisonicotinic acid 97%
2-Bromo-5-fluoroisonicotinic acid
Precaución: for Research Use Only
Formula:
C₆H₃BrFNO₂ MW: 219.9959 g/mol Boiling Pt: 406.7 °C (760 mm Hg) Flash Pt: 199.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09261246 CAS Number: 885588-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...