2-Bromo-5-fluoro-3-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A114496-100G
A114496-10G
A114496-250MG
A114496-25G
A114496-1G
A114496-5G
103798-416EA
8
USD
103798-416
103824-254
77257-834
77257-836
77257-838
77257-840
77311-064
77311-066
2-Bromo-5-fluoro-3-methylpyridine 98%
2-Bromo-5-fluoro-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 200.2 °C (760 mm Hg) Flash Pt: 96.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09835355 CAS Number: 38186-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...