2-Bromo-1,3-dimethyl-5-fluorobenzene 98%

Supplier: Ambeed
Warning

A248668-100G A248668-10G A248668-1G A248668-25G
103811-834EA 6 USD
103811-834 103851-668 76815-600 76815-602 77304-454 77304-456
2-Bromo-1,3-dimethyl-5-fluorobenzene 98%
2-Bromo-1,3-dimethyl-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
Boiling Pt: 206 °C (760 mm Hg)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD09864702
CAS Number: 14659-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR