2-Bromo-5-ethoxypyridine 95%

Supplier: Ambeed
Warning

A491913-1G A491913-250MG
103828-222EA 23 USD
103828-222 103839-296 76843-260 77265-594
2-Bromo-5-ethoxypyridine 95%
2-Bromo-5-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Boiling Pt: 256.062 °C (760 mm Hg)
Flash Pt: 108.663 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260481
CAS Number: 42834-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR