2-Bromo-5-cyanobenzoic acid 97%

Supplier: Ambeed
Warning

A153788-100MG A153788-250MG A153788-1G A153788-5G
103804-570EA 156 USD
103804-570 103842-748 76823-908 76823-910 77318-430 77318-432
2-Bromo-5-cyanobenzoic acid 97%
2-Bromo-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MW: 226.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847005
CAS Number: 845616-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR