2-Bromo-5-cyano-3-picoline 97%

Supplier: Ambeed
Warning

A135986-100MG A135986-5G
103802-254EA 59 USD
103802-254 77109-526 77298-836
2-Bromo-5-cyano-3-picoline 97%
2-Bromo-5-cyano-3-picoline

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
Boiling Pt: 291.5 °C (760 mm Hg)
Flash Pt: 130.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095063
CAS Number: 374633-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR