2-Bromo-5-chlorothiophene 98%

Supplier: Ambeed
Warning

A786344-1G A786344-25G A786344-100G
103840-996EA 17 USD
103840-996 76820-492 76845-248 77674-144
2-Bromo-5-chlorothiophene 98%
2-Bromo-5-chlorothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 176.5 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014523
CAS Number: 2873-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR