2-Bromo-5-chloropyrimidine 97%

Supplier: Ambeed
Warning

A187938-100MG A187938-10G A187938-250MG A187938-1G A187938-25G
103816-056EA 39 USD
103816-056 103846-648 76784-832 76784-834 76784-836 77084-366 77084-368
2-Bromo-5-chloropyrimidine 97%
2-Bromo-5-chloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
Boiling Pt: 291 °C (760 mm Hg)
Flash Pt: 130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12026299
CAS Number: 124405-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR