2-Bromo-5-chloropyrazine 97%

Supplier: Ambeed
Warning

A225706-10G A225706-1G A225706-25G A225706-5G A225706-100G
103819-190EA 8 USD
103819-190 103850-090 77030-940 77088-790 77088-792 77088-794 77667-874
2-Bromo-5-chloropyrazine 97%
2-Bromo-5-chloropyrazine

Precaución: for Research Use Only

Formula: C₄H₂BrClN₂
MW: 193.4291 g/mol
Boiling Pt: 208 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08460074
CAS Number: 912773-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR