2-Bromo-5-chlorophenol 97%

Supplier: Ambeed
Warning

A259787-25G A259787-5G A259787-100G A259787-10G A259787-1G
103812-464EA 8 USD
103812-464 103852-346 77091-928 77091-930 77262-180 77262-182 77304-776
2-Bromo-5-chlorophenol 97%
2-Bromo-5-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 222.1 °C (760 mm Hg)
Flash Pt: 88.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00142870
CAS Number: 13659-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR