2-Bromo-5-chloroisonicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-chloro-isonicotinic acid

A247162-1G A247162-250MG
103811-752EA 29 USD
103811-752 103851-584 77090-898 77672-208
2-Bromo-5-chloroisonicotinic acid 95%
2-Bromo-5-chloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.451 g/mol
Boiling Pt: 431.6 °C (760 mm Hg)
Flash Pt: 214.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13181649
CAS Number: 530156-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR