2-Bromo-5-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A101141-100G A101141-10G A101141-1G A101141-25G A101141-500G A101141-5G
103821-508EA 6 USD
103821-508 77107-192 77107-194 77107-196 77107-198 77107-200 77296-994
2-Bromo-5-chlorobenzoic acid 98%
2-Bromo-5-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
Boiling Pt: 326.5 °C (760 mm Hg)
Melting Pt: 153…157 °C
Flash Pt: 151.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00013982
CAS Number: 21739-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR