2-Bromo-5-chlorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A234850-100MG A234850-250MG A234850-1G
103810-982EA 37 USD
103810-982 103850-740 77031-152 77031-154 77304-002
2-Bromo-5-chlorobenzo[d]thiazole 97%
2-Bromo-5-chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MW: 248.52 g/mol
Boiling Pt: 330.5 °C (760 mm Hg)
Flash Pt: 153.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09749223
CAS Number: 2941-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR