2-Bromo-5-chlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A180142-1G A180142-25G A180142-5G A180142-100G A180142-10G A180142-500G
103807-516EA 5 USD
103807-516 103845-870 77083-358 77083-360 77083-362 77096-804 77096-806 77096-808
2-Bromo-5-chlorobenzaldehyde 98%
2-Bromo-5-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
Boiling Pt: 257.7 °C (760 mm Hg)
Melting Pt: 66 °C
Flash Pt: 109.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00462870
CAS Number: 174265-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR