2-Bromo-5-chloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A127009-10G A127009-25G A127009-5G A127009-1G
103800-898EA 6 USD
103800-898 76810-968 76810-970 77298-164 77298-166 77313-146
2-Bromo-5-chloro-3-methylpyridine 98%
2-Bromo-5-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Melting Pt: 40…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095062
CAS Number: 65550-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR