2-Bromo-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A479676-25G A479676-100G A479676-10G A479676-500G A479676-5G
103839-190EA 10 USD
103839-190 76843-132 77156-422 77156-424 77156-426 77307-530
2-Bromo-5-(trifluoromethyl)pyridine 98%
2-Bromo-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 209.3 °C (760 mm Hg)
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR