2-Bromo-5-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A119877-100G A119877-10G A119877-250MG A119877-25G A119877-1G A119877-5G
103799-852EA 10 USD
103799-852 77258-058 77258-060 77258-062 77258-064 77311-960 77311-962
2-Bromo-5-(trifluoromethyl)benzoic acid 97%
2-Bromo-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.0154 g/mol
Boiling Pt: 301 °C (760 mm Hg)
Melting Pt: 116…117 °C
Flash Pt: 135.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06208253
CAS Number: 1483-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR