2-Bromo-5-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A276886-10G A276886-25G A276886-5G
103813-324EA 9 USD
103813-324 103853-260 76772-248 77093-124 77093-126
2-Bromo-5-(trifluoromethyl)benzaldehyde 97%
2-Bromo-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
Boiling Pt: 239.1 °C (760 mm Hg)
Flash Pt: 136.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973760
CAS Number: 102684-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR