2-Bromo-5-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-bromobenzotrifluoride, 6-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-4-bromo-α,α,α-trifluorotoluene, 3-Amino-4-brom-α,α,α-trifluortoluol

A179524-10G A179524-500G A179524-25G A179524-5G
103845-788EA 6 USD
103845-788 76769-520 76769-522 77083-256 77083-258
2-Bromo-5-(trifluoromethyl)aniline 98%
2-Bromo-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 243.1 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007637
CAS Number: 454-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR