2-Bromo-5-(trifluoromethoxy)pyridine 96%

Supplier: Ambeed
Danger

A122369-100MG A122369-10G A122369-250MG A122369-5G
103800-212EA 90 USD
103800-212 76781-824 76781-826 76781-828 77312-346
2-Bromo-5-(trifluoromethoxy)pyridine 96%
2-Bromo-5-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃NO
MW: 241.993 g/mol
Boiling Pt: 194.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10698596
CAS Number: 888327-36-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR