2-Bromo-5-(trifluoromethoxy)benzoic acid 96%

Supplier: Ambeed
Warning

A445081-1G A445081-100MG A445081-250MG
103827-726EA 34 USD
103827-726 103838-790 76842-570 76990-658 76990-660
2-Bromo-5-(trifluoromethoxy)benzoic acid 96%
2-Bromo-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₃
MW: 285.015 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Flash Pt: 129.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07368726
CAS Number: 403646-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR