2-Bromo-5-(hydroxymethyl)phenol 97%
Supplier: Ambeed
|
Warning
|
A391212-1G
A391212-25G
A391212-100MG
A391212-250MG
103826-874EA
12
USD
103826-874
103837-914
76841-302
76841-304
76876-778
76876-780
2-Bromo-5-(hydroxymethyl)phenol 97%
2-Bromo-5-(hydroxymethyl)phenol
Precaución: for Research Use Only
Formula:
C₇H₇BrO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD14705103 CAS Number: 2737-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...