2-Bromo-5-(hydroxymethyl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
A174836-1G
A174836-100MG
A174836-250MG
103806-978EA
304
USD
103806-978
103845-278
77082-584
77163-680
77163-682
2-Bromo-5-(hydroxymethyl)benzonitrile 98%
2-Bromo-5-(hydroxymethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆BrNO Boiling Pt: 360.8±32.0 °C (760 mm Hg) Flash Pt: 172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18390752 CAS Number: 1261609-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...